Learn More
Thermo Scientific™ Three60™ Sampling System

Description
The Thermo Scientific™ Three60™ Sampling System allows you to take a representative sample of your product with minimal effort. The fluid paths of the four pre-irradiated BioProcess Containers (BPCs) and assemblies feature quick-turn valves and pinch-and-cut disconnections for fluid transfer and removal. The Three60 Sampling System is composed of Thermo Scientific™ ASI™ 77 film, a 5-layer, 7mL, polyethylene film and includes 2 ports (1 luer lock insert + cap, 1 injection site).
- Compatible with a 3.8 cm (1.5-inch) sanitary connector
- Requires no expensive hardware
- Each Three60 package contains a valve and four assemblies with pinch-and-cut disconnections
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
Applications
- Removes the need for tank cleaning and helps to reduce cycle times
- Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Content And Storage | Store at room temperature. |
| Dimensions | 22.0 x 13.5 cm (8.66 x 5.32 in.) |
| Film Description | ASI™ 77 |
| Outer Packaging | Placed into a thermoform tray and sealed |
| Port Detail | 2 Ports (1 Luer Lock Insert + Cap, 1 Injection Site) |
| Material | Ethyl Vinyl Acetate |
| Quantity | 8/Cs. |
| Shipping Condition | Room Temperature |
| Sterilization Method | Irradiated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.