Learn More
Abcam Threonine Assay Kit, Fluorescent, 100 Tests
Description
- Detection method: Fluorescent
- Sample types: Tissue Homogenate, Cell Lysate, Serum, Plasma, Cerebral Spinal Fluid
- Equipment: Microplate reader
- Citations: Cited in > 2 publications
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Threonine |
| No. of Reactions | 100 Tests |
| For Use With (Equipment) | Microplate Reader |
| Detection Location | In-Solution Detection |
| Format | Kit |
| Detection Method | Fluorescence |
| Sample Type | Tissue Homogenate, Cell Lysate, Serum, Plasma, Cerebral Spinal Fluid |
| Quantity | 100 Tests |
| For Use With (Application) | Threonine detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.