Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Threonine, Crystal, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18607387 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
5 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-387 5 g Amber Glass Bottle
18-607-390 500 g Poly Bottle
18-607-392 5 kg Poly Pail
18-607-393 50 kg Fiber Drum
18-607-388 25 g Poly Bottle
18-607-389 100 g Poly Bottle
18-607-391 1 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-387 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T10565GM
Add to Cart
Add to Cart

T1056, 72-19-5, C4H9NO3

Spectrum™ Chemical Threonine, Crystal, USP is an antispacitiy agent used to treat various nervous system disorders including spinal spasticity, multiple sclerosis, familial spastic paraparesis, and amyotrophic lateral sclerosis disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
IUPAC Name 2-amino-3-hydroxybutanoic acid
SMILES CC(O)C(N)C(O)=O

Specifications

CAS Max % 1
pH 5.0 to 6.5
Ignition Residue 0.004
Loss on Drying 0.002
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation -26.7° to -29.1°
Percent Purity 98.5 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.