Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombin Monoclonal Antibody (5020)
GREENER_CHOICE

Catalog No. MA143019
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA143019 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA143019 Supplier Invitrogen™ Supplier No. MA143019
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-43019 detects Thrombin in human samples. MA1-43019 has successfully been used in ELISA applications. MA1-43019 is specific to human thrombin, thrombin-ATIII complex and thrombin-PPACK.

Coagulation factor II is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombin
Applications ELISA
Classification Monoclonal
Clone 5020
Concentration 8.2 mg/mL
Conjugate Unconjugated
Formulation 50% water with 50% glycerol and no preservative
Gene F2
Gene Accession No. P00734
Gene Alias Activation peptide fragment 1; Activation peptide fragment 2; Cf2; Cf-2; coagulation factor 2; coagulation factor II; coagulation factor II (thrombin); coagulation factor II, thrombin; F2; FII; prepro-coagulation factor II; Prothombin; prothrombin; prothrombin B-chain; PT; RPRGL2; serine protease; THPH1; thrombin; Thrombin heavy chain; Thrombin light chain
Gene Symbols F2
Host Species Mouse
Immunogen Human Thrombin.
Purification Method SDS-PAGE
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2147
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.