Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombin, MP Biomedicals

Catalog No. ICN15416350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50kU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15416350 50kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15416350 Supplier MP Biomedicals, Inc Supplier No. 0215416350
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thrombin is prepared from fresh bovine plasma by an ion exchange method.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-04-4

Specifications

Grade High Purity
Content And Storage Store at -20°C
Quantity 50kU
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.