Learn More
CPC Scientific H-Ala-Phe-Leu-Ala-Arg-Ala-Ala-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific THRO007A
Encompass
This is a peptide analogue of thrombin receptor activating peptide. It is been shown to block alpha-thrombin and thrombin receptor activating peptide-induced platelet aggregation in vitro and inhibit thrombosis in in vivo animal models.SEQUENCE: H-Ala-Phe-Leu-Ala-Arg-Ala-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: AFLARAAMOLECULAR FORMULA: MOLECULAR WEIGHT: 718.9STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: HematologyREFERENCES:Pakala, R. et al. Thromb Res. 100, 89 (2000)Pakala, R. et al. J. of Cardiovascular Pharm. 37, 619 (2001)SKU(s): THRO-007A
Catalog No. 50-211-0570
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.