Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pacific Hemostasis™ Thromboplastin-DS, 10 x 4mL

Detect acquired or congenital disorders affecting the extrinsic and common pathways.

Catalog No. 292273

This product cannot be ordered online due to special handling requirements. Please contact National Order Desk customer service at CS.NOD@thermofisher.com or 1-800-361-9402 for ordering assistance.

This item is not returnable. View return policy

Description

Description

Detect acquired or congenital disorders affecting the extrinsic and common pathways with Thermo Scientific™ Pacific Hemostasis™ Prothrombin Time (PT) Reagents. Widely accepted as a means of monitoring patients on oral anticoagulant therapy such as Warfarin/Coumadin, reagents are manufactured with up to three years extended shelf life and have excellent lot-to-lot consistency.

Features:
  • Compatible with automated, semi-automated or manual systems
  • Reconstitution stability: 7 days at 2° to 8°C (36° to 46°F)
  • Dried rabbit brain thromboplastin

Thromboplastin-D with CaCl2 (Product No. 10-0356, 10-0357):

  • Target ISI value of 1.7 to 2.2
  • Expected normal values of 12 to 15 seconds

Thromboplastin-DS (Product No. 10-0362, 10-0354):

  • Target ISI value of 1.0 to 1.3
  • Expected normal values of 12 to 15 seconds
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.