Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Thrombopoietin ELISA Kit

Catalog No. EHTHPOX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHTHPOX5 5 x 96 Tests
EHTHPO 96 Tests
EHTHPOX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHTHPOX5 Supplier Invitrogen™ Supplier No. EHTHPOX5
Add to Cart
Add to Cart

ELISA

The Human Thrombopoietin (TPO) ELISA quantitates Hu TPO in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu TPO. Principle of the method The Human TPO solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Thrombopoietin (TPO, THPO, MPL ligand, megakaryocyte colony-stimulating factor) is responsible for the generation of platelets through its action on hematopoietic cells committed to the megakaryocyte lineage. Binding of thrombopoietin to its receptor, MPL (CD110), initiates a signaling cascade via JAK/STAT, MAPK, and PI3K/AKT signaling pathways that leads to the proliferation and differentiation of megakaryocytes and, ultimately, to the production of platelets (thrombopoiesis). Thrombopoietin is constitutively produced by the liver and has a predicted molecular weight of 38 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40225
Assay Range 93.75 to 6000 pg/mL
Assay Sensitivity 90 pg/mL
Calibration Range 90 pg/mL to 6000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Growth Factors & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the following cytokines tested: Human BDNF, BLC, ENA-78, FGF-4, IL- 1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-9, IL-11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, G-CSF, GM-CSF, IFN-gamma, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PDGF, RANTES, SCF, TARC, TGF-beta1, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, VEGF.
Sufficient For 5 x 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 7066
Gene Symbol MGDF,MKCSF,ML,MPLLG,THCYT1,TPO
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Cell Cycle & Proliferation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym MGDF, MKCSF, ML, MPLLG, THCYT1, TPO
Target Thrombopoietin
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.