Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 1 Monoclonal Antibody (A6.1)
GREENER_CHOICE

Catalog No. MA513398
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA513398 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA513398 Supplier Invitrogen™ Supplier No. MA513398
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA5-13398 targets Thrombospondin in FACS, ICC/IF, IHC (P), IM, IP, and WB applications and shows reactivity with Bovine, Canine, Equine, Human, mouse, Ovine, Porcine, and Rat samples. The MA5-13398 immunogen is reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunomicroscopy, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene THBS1
Gene Accession No. P07996, P35441, Q28178
Gene Alias glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen Reduced and alkylated purified human TSP (fully denatured) from the supernatant of thrombin-activated platelets.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100057478, 101106433, 21825, 281530, 445442, 487486, 492313, 7057
Target Species Bovine, Canine, Horse, Human, Mouse, Sheep, Pig, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.