Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin-1 Monoclonal Antibody (A6.1), eBioscience™
GREENER_CHOICE

Catalog No. 501122826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-2826 100 μg
14-975-680 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-2826 Supplier Invitrogen™ Supplier No. 14975682
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The A6.1 monoclonal antibody recognizes human, mouse, and rat thrombospondin-1, a member of the thrombospondin family of extracellular calcium-binding proteins. Thrombospondin-1 is a secreted glycoprotein that was first discovered in activated platelets but is also expressed by endothelial cells, adipocytes, fibroblasts, macrophages, monocytes, and smooth muscle cells. Thrombospondin-1 functions by binding members of the extracellular matrix such as fibronectin, integrins, and heparin, and its signaling is mediated through activation of the TGF beta signaling pathway. Thrombospondin-1 has been shown to be involved in angiogenesis, inflammation, osteogenesis, cell proliferation, and synaptogenesis during neural development. Applications Reported: This A6.1 antibody has been reported for use in immunoprecipitation, western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, ELISA, and immunocytochemistry. Applications Tested: This A6.1 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using high pH antigen retrieval and can be used at less than or equal to 1 μg/mL. This A6.1 antibody has been tested by immunocytochemistry of formaldehyde or methanol fixed and permeabilized human cells and can be used at less than or equal to 1 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater th...

Thrombospondin is a protein from platelet a-granules. It is made up of three identical subunits bound by interchain disulfides. It is secreted at sites of platelet activation and aggregation and is involved in the processes of chemotaxis, adhesion, proliferation and differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin-1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone A6.1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene THBS1
Gene Accession No. P07996, P35441
Gene Alias glycoprotein G; tbsp1; THBS; Thbs1; Thbs-1; thrombospondin; thrombospondin 1; thrombospondin-1; thrombospondin-1p180; thrombospondin-p50; ThrombospondinTHBS1; TSP; TSP1; TSP-1
Gene Symbols THBS1
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21825, 445442, 7057
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.