Learn More
Diagnostic Biosystems THROMBOSPONDIN Caspase 3
Supplier:  Diagnostic Biosystems MOB30905
Encompass_Preferred
This antibody is specific to 450 kDa (non-reduced) and 170 to 180 kD (reduced) form of thrombospondin. Thrombospondin is a protein from platelet α granules. It is secreted at sites of platelet activation and aggregation and is involved in the differentiation of leukocytes, fibroblasts, smooth muscle and endothelial cells.
Catalog No. 50-238-5714
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.