Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thrombospondin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580123 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580123 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580123 Supplier Invitrogen™ Supplier No. PA580123
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, human U87 whole cell, human U251 whole cell.

The Thrombospondin proteins (TSP 1-5) compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and hetero-multimers. Thrombospondins play a role in development, aggregation of platelets, adhesion and migration of cells and progression of cells through the growth cycle. Thrombospondin 1 is released from platelets in response to Thrombin stimulation and is a transient component of the extracellular matrix of developing and repairing tissues. Thrombospondin 2 shares a high degree of homology with Thrombospondin 1, and is thought to have overlapping but unique functions. Thrombospondin 3 is a developmentally regulated heparin-binding protein. Thrombospondin 4 is neuronally expressed and stimulates neurite outgrowth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin 2
Applications Western Blot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation Antibody with 0.2mg Na2PO4, 0.9mg NaCl, 4mg trehalose and no preservative
Gene THBS2
Gene Accession No. P35442
Gene Alias Thbs2; Thbs-2; thrombospondin 2; Thrombospondin2; thrombospondin-2; TSP2; TSP-2; TSP2thrombospondin-2
Gene Symbols THBS2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human Thrombospondin 2 (637-655aa EAAKTEKQVCEPENPCKDK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7058
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.