Learn More
Cayman Chemical Thromboxane B2 Express ELISA Kit - Monoclonal, 96 solid wells
SDP
Supplier:  Cayman Chemical 1000402396SOLIDWELLS
Encompass_Preferred
Thromboxane A2 (TXA2), produced from arachidonic acid, causes irreversible platelet aggregation and contraction of vascular and bronchial smooth muscle. TXA2 is rapidly hydrolyzed non-enzymatically to TXB2, which is then quickly metabolized to urinary metabolites for clearance by the kidneys. Urinary analysis of TXB2 accurately reflects intrarenal TXA2 synthesis, while measurement of 11-dehydro and 2,3-dinor TX metabolites gives the best estimate of systemic TXA2 secretion. Cayman’s TXB2 Express ELISA is a competitive assay that provides a rapid, accurate measurement of TXB2 from plasma, urine, and cell culture supernatants. The assay has a range from 15.6-2,000 pg/ml and a sensitivity (80% B/B0) of approximately 45 pg/ml.
Catalog No. NC0190287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.