Learn More
Fisherbrand™ Thumb Cap
Catalog No. 23001709 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Green
Red
White
Closure Size:
12/13 mm
13 mm
16 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Size
23-001-709 Green 13 mm
23-001-710 Red 16 mm
23-001-834 Green 12/13 mm
23-001-835 White 13 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23-001-709
Add to Cart
Add to Cart
Colored thumb cap with dual tab.
  • Features a dual tab
  • Available in various colors and sizes
  • 1000 per case
Specifications
Liner Material Unlined
Closure Size 13 mm
Autoclavable Not Autoclavable
Material Plastic
Color Green
No. per Case 1000
Sterility Non-sterile
Disposable Single-use
Fit for Purpose PCR Ready