Learn More
Description
Specifications
Specifications
| Description | Antibody Labeling Kit |
| Target | Thunder-Link PLUS oligo |
| For Use With (Application) | Warm the Thunder-Link PLUS Conjugate Clean Up Reagent by placing the tube in warm water (not warmer than 40C) for 10 minutes, then shaking to re-dissolve the contents. Once dissolved, maintain the tube at ∼22C to prevent any crystal formation before use. |
| Content And Storage | Store at 4°C. Do not freeze. |
| Product Line | Novus |
| Quantity | 300 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
