Learn More
Cayman Chemical THUNDERHumn IL1 TRFRET 480
SDP
Supplier:  Cayman Chemical 500230480 WELL
Encompass_Preferred
A PPARY inverse agonist acts as an inverse agonist in a co-repressor reporter assay with an NCOR2 peptide (EC50 5.8 nM) selective for PPARY (IC50s 0.14 and 0.4 nM for the mouse and human receptors respectively) over PPARa and PPAR in reporter assays using CHO cells (IC50 >50 UM for both human receptors) inhibits the proliferation of UM-UC-9 cells (IC50 3.4 nM)
Catalog No. 50-289-6967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.