Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Thymidine-5'-monophosphate disodium salt

Catalog No. AAJ6074703
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6074703 1 g
AAJ60747MC 100 mg
AAJ60747MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6074703 Supplier Thermo Scientific Chemicals Supplier No. J6074703
Add to Cart

CAS: 33430-62-5 | C10H13N2Na2O8P | 366.17 g/mol

Thymidine-5'-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine-5′-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

Solubility
Soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33430-62-5
Molecular Formula C10H13N2Na2O8P
Molecular Weight (g/mol) 366.17
MDL Number MFCD00023797
InChI Key AGSQMPPRYZYDFV-ZJWYQBPBSA-L
Synonym tmp disodium
PubChem CID 131674985
IUPAC Name disodium;[(2R,3S,5R)-3-hydroxy-5-(5-methyl-4-oxido-2-oxopyrimidin-1-yl)oxolan-2-yl]methyl hydrogen phosphate;hydrate
SMILES [Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O

Specifications

Color White
Quantity 1 g
Beilstein 4086927
Solubility Information Soluble in water.
Formula Weight 366.18
Physical Form Powder
Chemical Name or Material Thymidine-5'-monophosphate disodium salt
EINECSNumber 251-518-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.