Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine, [Methyl-3H], MP Biomedicals™

Catalog No. MP012404201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mci; 37 mbq
250 μci; 9.25 mbq
5 mci; 185 mbq
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP012404201 1 mci; 37 mbq
MP01240422 250 μci; 9.25 mbq
MP012404205 5 mci; 185 mbq
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP012404201 Supplier MP Biomedicals, Inc Supplier No. 012404201

Molecular Formula C10H14N2O5

  • Sp. Act. 40-60 Ci/mmol; 1.48-2.22 TBq/mmol
  • Sterile aqueous solution.
  • HPLC Analyzed for Purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10H14N2O5

Specifications

Quantity 1 mci; 37 mbq
storageNote1 Store at 2-8°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.