Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymidine, MP Biomedicals™

Catalog No. MP219475405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219475405 5 g
MP219475401 1 g
MP219475410 10 g
MP219475425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP219475405 Supplier MP Biomedicals, Inc Supplier No. 0219475405
Add to Cart
Add to Cart

Used in forming hybrid cell lines where cells are incubated in HAT medium. Thymidine, MP Biomedicals, a cell culture reagent, is a nucleoside composed of deoxyribose (a pentose sugar) joined to the pyrimidine base thymine. It is highly stable and appears

Mainly used in forming hybrid cell lines using HAT (hypoxanthine-aminopterin-thymidine) medium to give rise to monoclonal antibodies. Also used in HT and HAT cocktails for hybridoma fusion, selection and cloning.

Deoxythymidine can be phosphorylated with one, two or three phosphoric acid groups, creating respectively dTMP, dTDP or dTTP (deoxythymidine mono-, di-, or triphosphate). The stability of deoxythymidine under standard temperature and pressure (STP) is very high. Thymidine is listed as a chemical teratogen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-89-5
Molecular Formula C10H14N2O5
Molecular Weight (g/mol) 242.23
InChI Key IQFYYKKMVGJFEH-UHFFFAOYNA-N
Synonym thymidine, deoxythymidine, 2'-deoxythymidine, 5-methyldeoxyuridine, thymidin, beta-thymidine, dthyd, 5-methyl-2'-deoxyuridine, thymine-2-deoxyriboside, thyminedeoxyriboside
PubChem CID 5789
ChEBI CHEBI:17748
IUPAC Name 1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES CC1=CN(C2CC(O)C(CO)O2)C(=O)NC1=O

Specifications

Melting Point 185°C to 191°C
Quantity 5 g
Solubility Information Soluble in water, methanol, hot alcohol, hot acetone, hot ethyl acetate pyridine, glacial acetic acid and HCl (20mg/mL, clear, colorless solution); sparingly soluble in hot chloroform.
Optical Rotation 17.5–19.5° (c=1, water)
Formula Weight 242.2
Percent Purity >99.9%
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.