Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thymidine Phosphorylase Monoclonal Antibody (826722)
GREENER_CHOICE

Catalog No. MA561844
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561844 Supplier Invitrogen™ Supplier No. MA561844
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymidine Phosphorylase
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 826722
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Gene TYMP
Gene Accession No. P19971, Q5FVR2, Q99N42
Gene Alias 2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP
Gene Symbols TYMP
Host Species Rat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse PD-ECGF/Thymidine Phosphorylase, Met1-Pro471.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1890, 315219, 72962
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.