Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thymidine Phosphorylase Monoclonal Antibody (PGF 44C)
GREENER_CHOICE

Catalog No. MA146059
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA146059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA146059 Supplier Invitrogen™ Supplier No. MA146059
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Suggested positive control: HUVEC cells, breast carcinoma.

A platelet-derived endothelial growth factor (PD-ECGF), same as thymidine phosphorylase (TP) or gliostatin. In the presence of inorganic orthophosphate, it catalyses the reversible phospholytic cleavage of thymidine and deoxyuridine to their corresponding bases and 2-deoxyribose-1-phosphate. It is both chemotactic and mitogenic for endothelial cells and a non-heparin binding angiogenic factor present in platelets. It is also involved in transformation of fluoropyrimidines, cytotoxic agents used in the treatment of a variety of malignancies, into active cytotoxic metabolites.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thymidine Phosphorylase
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone PGF 44C
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TYMP
Gene Accession No. P19971
Gene Alias 2900072D10Rik; ECGF; Ecgf1; endothelial cell growth factor 1 (platelet-derived); gliostatin; hPD-ECGF; MEDPS1; MNGIE; MTDPS1; OTTHUMP00000196767; OTTHUMP00000196770; PD TP; PDECGF; PD-ECGF; Pdgfec; platelet derived growth factor, endothelial cell; Platelet-derived endothelial cell growth factor; TdRPase; thymidine phosphorylase; TP; TYMP
Gene Symbols TYMP
Host Species Mouse
Immunogen Full length human protein in E. coli.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1890
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.