Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymine, 97%

Catalog No. AAA1587914
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587914 25 g
AAA1587922 100 g
AAA1587936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1587914 Supplier Thermo Scientific Chemicals Supplier No. A1587914
Add to Cart

CAS: 65-71-4 | C5H6N2O2 | 126.115 g/mol

Thymine is one of the four nucleobases, along with adenine, guanine and cytosine found in deoxyribonucleic acids (DNA). Thymine may be used to study chemical processes that affect DNA structure, such a radiation induced radical production leading to base cross-linking reactions and derivitizations. Thymine may be used to study the parameters of hydrogen bonding kinetics and energies with other nucleobases such as adenine. Thymine is used to develop sensitive heavy metal (mercury) detectors based on coordination chemistry and nanoparticle structures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymine is one of the four nucleobases, along with adenine, guanine and cytosine found in deoxyribonucleic acids (DNA). Thymine may be used to study chemical processes that affect DNA structure, such a radiation induced radical production leading to base cross-linking reactions and derivitizations. Thymine may be used to study the parameters of hydrogen bonding kinetics and energies with other nucleobases such as adenine. Thymine is used to develop sensitive heavy metal (mercury) detectors based on coordination chemistry and nanoparticle structures.

Solubility
Soluble in hot water. Slightly soluble in alcohol.

Notes
Store in cool dry place.. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-71-4
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.115
MDL Number MFCD00006026
InChI Key RWQNBRDOKXIBIV-UHFFFAOYSA-N
Synonym thymine, 5-methyluracil, thymin, 2,4-dihydroxy-5-methylpyrimidine, thymine anhydrate, 2,4 1h,3h-pyrimidinedione, 5-methyl, 5-methylpyrimidine-2,4 1h,3h-dione, 5-methyl-2,4 1h,3h-pyrimidinedione, thymin purine base, 5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione
PubChem CID 1135
ChEBI CHEBI:17821
IUPAC Name 5-methyl-1H-pyrimidine-2,4-dione
SMILES CC1=CNC(=O)NC1=O

Specifications

Melting Point 316°C to 317°C
Assay Percent Range 97%
Quantity 25 g
Beilstein 607626
Merck Index 14,9398
Solubility Information Soluble in hot water. Slightly soluble in alcohol.
Formula Weight 126.12
Percent Purity 97%
Chemical Name or Material Thymine
Hazard Category H319-H500
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 200-616-1
RTECSNumber XP2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.