Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Thymol
SCI_ED

Catalog No. S25603 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25603 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25603
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
Synonym thymol, 2-isopropyl-5-methylphenol, 5-methyl-2-isopropylphenol, thyme camphor, 3-p-cymenol, 6-isopropyl-m-cresol, thymic acid, isopropyl cresol, 5-methyl-2-1-methylethyl phenol, 6-isopropyl-3-methylphenol
PubChem CID 6989
ChEBI CHEBI:27607
IUPAC Name 5-methyl-2-propan-2-ylphenol
SMILES CC1=CC(=C(C=C1)C(C)C)O

Specifications

Density 0.969g/mL
Assay Percent Range 99 to 100%
Packaging Lab Grade
Linear Formula CH3(C3H7)C6H3OH
Quantity 100 g
Formula Weight 150.22
Grade Laboratory
Chemical Name or Material Thymol
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.