CAS RN 89-83-8
Thymol, 99%
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 MDL Number: MFCD00002309 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N Synonym: thymol,2-isopropyl-5-methylphenol,5-methyl-2-isopropylphenol,thyme camphor,3-p-cymenol,6-isopropyl-m-cresol,thymic acid,isopropyl cresol,5-methyl-2-1-methylethyl phenol,6-isopropyl-3-methylphenol PubChem CID: 6989 ChEBI: CHEBI:27607 IUPAC Name: 5-methyl-2-propan-2-ylphenol SMILES: CC1=CC(=C(C=C1)C(C)C)O
Thymol, 98+%
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.221 MDL Number: MFCD00002309 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N Synonym: 2-Isopropyl-5-methylphenol PubChem CID: 6989 ChEBI: CHEBI:27607 IUPAC Name: 5-methyl-2-propan-2-ylphenol SMILES: CC1=CC(=C(C=C1)C(C)C)O
Thymol Standard, For quantitative NMR, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Thymol, Sigma-Aldrich
CAS: 89-83-8
Thymol 99.0+%, TCI America™
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.221 MDL Number: MFCD00002309 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N Synonym: thymol,2-isopropyl-5-methylphenol,5-methyl-2-isopropylphenol,thyme camphor,3-p-cymenol,6-isopropyl-m-cresol,thymic acid,isopropyl cresol,5-methyl-2-1-methylethyl phenol,6-isopropyl-3-methylphenol PubChem CID: 6989 ChEBI: CHEBI:27607 IUPAC Name: 5-methyl-2-propan-2-ylphenol SMILES: CC1=CC(=C(C=C1)C(C)C)O
Standard Hard Water, 200 ppm Hardness as CaCO3, Ricca Chemical
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.221 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N PubChem CID: 6989 ChEBI: CHEBI:27607 IUPAC Name: 5-methyl-2-propan-2-ylphenol SMILES: CC1=CC(=C(C=C1)C(C)C)O
Thymol, Purified, Reagents
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N Synonym: Thyme camphor, 2-Isopropyl-5-methylphenol, 5-Methyl-2-isopropylphenol IUPAC Name: 5-methyl-2-(propan-2-yl)phenol SMILES: CC(C)C1=CC=C(C)C=C1O
Medisca Thymol NF (Crystals)
Thymol NF comes as colorless, often large, crystals, or white, crystalline powder. Has an aromatic, thyme-like odor. It is hygroscopic and is affected by light.
Fisher Science Education™ Thymol
CAS: 89-83-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.221 InChI Key: MGSRCZKZVOBKFT-UHFFFAOYSA-N Synonym: thymol,2-isopropyl-5-methylphenol,5-methyl-2-isopropylphenol,thyme camphor,3-p-cymenol,6-isopropyl-m-cresol,thymic acid,isopropyl cresol,5-methyl-2-1-methylethyl phenol,6-isopropyl-3-methylphenol PubChem CID: 6989 ChEBI: CHEBI:27607 IUPAC Name: 5-methyl-2-propan-2-ylphenol SMILES: CC1=CC(=C(C=C1)C(C)C)O