Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Blue, 0.08% in Methanol, for Organic Acids, Certified, LabChem™

Catalog No. LC260501 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC260501 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC260501 Supplier LabChem, Inc. Supplier No. LC260501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76-16-9 , 67-56-1 , 1273376-61-6

Specifications

Melting Point -98°C
Boiling Point 65°C
CAS Max % 99.92, 0.08
Linear Formula C27H29NaO5S
UN Number UN1993
Solubility Information Soluble in water
Identification Passes Test
Formula Weight 488.58
Grade Certified
Packaging Poly Bottle
Specific Gravity 0.70g/mL
Decomposition Information Sulfur compounds; Carbon monoxide; Carbon dioxide; May release flammable gases
Density 0.70g/mL
Color Red
Quantity 500 mL
Flash Point 9.7°C
Physical Form Liquid
Chemical Name or Material Thymol Blue, 0.08% in Methanol, for Organic Acids
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Toxic if swallowed, in contact with skin or if inhaled.
Causes damage to organs (central nervous system, kidneys, liver, optic nerve).
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Refrigerate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.