Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol Blue Indicator, 0.04% (w/v), Ricca Chemical
SDP

Catalog No. 840016 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8400-16 500 mL Natural Poly Bottle
8400-4 120 mL Natural Poly Bottle
R84000001A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 8400-16 Supplier Ricca Chemical Company Supplier No. 840016
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62625-21-2 , 7732-18-5
Molecular Formula C27H29NaO5S
Molecular Weight (g/mol) Mixture
MDL Number MFCD00151093
InChI Key OCMIKNBSIRSUPI-FRWNMSGJSA-M
PubChem CID 23692293
IUPAC Name sodium 2-{[4-hydroxy-2-methyl-5-(propan-2-yl)phenyl][(1E)-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene]methyl}benzene-1-sulfonate
SMILES [Na+].CC(C)C1=C\C(=C(\C2=CC(C(C)C)=C(O)C=C2C)C2=C(C=CC=C2)S([O-])(=O)=O)C(C)=CC1=O

Specifications

CAS Min % 98.0, 0.04
CAS Max % 100.0, 0.04
Concentration 0.04% (w/v)
Percent Purity 0.04% W/V
Grade Indicator
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Color Dark Green
pH 9
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Thymol Blue Indicator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.