Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Thymol Blue, pH Indicator, Solid, 25 g

Catalog No. 502704756
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4756 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4756 Supplier Abcam Supplier No. AB14629125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH indicator with visual transition intervals of pH 1.2 to 2.8 and pH 8.0 to 9.2.

  • Molecular weight: 466.6 g/mol
  • Form: Solid
  • pH indicator visual transition intervals: pH 1.2 (red) to pH 2.8 (yellow), and pH 8.0 (yellow) to pH 9.2 (blue)
  • Composition: 100% Thymol blue

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at ambient temperature.
Form Solid
Shipping Condition Ambient
Detection Method Colorimetric
Quantity 25 g
For Use With (Application) pH measurement
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.