Learn More
Description
Thymol NF comes as colorless, often large, crystals, or white, crystalline powder. Has an aromatic, thyme-like odor. It is hygroscopic and is affected by light.
Alternative name(s): 2-Isopropyl-5-methylphenol, 3-Hydroxy-para-cymene, 6-Isopropyl-m-cresol, alpha-Cymophenol, Thyme camphor
Specifications
Specifications
| Chemical Name or Material | Thymol NF (Crystals) |
| Packaging | Bottle, 60mL, Amber Glass, Wide Mouth Packer, 33-400 |
| CAS | 89-83-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 150.2196 |
| Quantity | 25 g |
| Percent Purity | 99 to 101 % |
| Grade | NF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
