Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Thymol NF (Crystals)
SDP

Catalog No. 14185468 Shop All Medisca Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-185-468 100 g
14-185-463 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-185-468 Supplier Medisca Supplier No. 030905
Add to Cart
Add to Cart

Thymol NF comes as colorless, often large, crystals, or white, crystalline powder. Has an aromatic, thyme-like odor. It is hygroscopic and is affected by light.

Thymol NF comes as colorless, often large, crystals, or white, crystalline powder. Has an aromatic, thyme-like odor. It is hygroscopic and is affected by light.

Alternative name(s): 2-Isopropyl-5-methylphenol, 3-Hydroxy-para-cymene, 6-Isopropyl-m-cresol, alpha-Cymophenol, Thyme camphor

Specifications

Chemical Name or Material Thymol NF (Crystals)
Packaging 300cc Amber Glass Bottle w/m
CAS 89-83-8
Type Drug Standard
Molecular Weight (g/mol) 150.2196
Quantity 100 g
Percent Purity 99 to 101 %
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.