Learn More
Abcam Thymol, Phenolic Disinfectant, >98% Purity, Solid, 25 g
Description
- Molecular weight: 150.22 g/mol
- Purity: >98%
- Form: Solid
- Constituents: 100% Thymol
- Applications: Used to inhibit oral bacteria, as a cosmetic biocide/preservative, fragrance ingredient, and denaturant.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Buffer | 100% Thymol |
| Shipping Condition | Ambient |
| Quantity | 25 g |
| For Use With (Application) | Inhibit oral bacteria, cosmetic biocide, preservative, fragrance ingredient, denaturant |
| Storage Requirements | Store at ambient temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.