Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol, Purified, Reagents

Catalog No. 01336931
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-931 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-931 Supplier Reagents Holdings Llc Supplier No. C233840025C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thymol, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 89-83-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
Synonym Thyme camphor, 2-Isopropyl-5-methylphenol, 5-Methyl-2-isopropylphenol
IUPAC Name 5-methyl-2-(propan-2-yl)phenol
SMILES CC(C)C1=CC=C(C)C=C1O

Specifications

Melting Point 49°C
Color Colorless/White
Melting Range 49°C
Packaging Glass Bottle
Quantity 25 g
Specific Gravity 0.9699
Grade Pure
Physical Form Solid
Chemical Name or Material Thymol, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.