Learn More
Thymol, Sigma-Aldrich
Catalog No. 01258948 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-948 100 g
01-258-805 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-948 Supplier MilliporeSigma Supelco Supplier No. T0501100G
Add to Cart
Add to Cart
Thymol is used as an additive in hematoxylin and eosin staining solutions to inhibit mold growth and as a reagent in antimicrobial and antifungal susceptibility testing.
Thymol is generally added in the Hematoxylin and Eosin staining solution to prevent mold growth. It is also suitable for use in antimicrobial screening by the disk diffusion method.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 89-83-8
Specifications
Quantity 100 g
Product Type Stain Kit
The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA's Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada's Pest Management Regulatory Agency, Turkey's Biocidal Products Regulation, Korea's Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.