Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymol, Sigma-Aldrich

Catalog No. 01258805 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-805 500 g
01-258-948 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-805 Supplier MilliporeSigma Supelco Supplier No. T0501500G
Add to Cart
Add to Cart

Thymol is generally added in the Hematoxylin and Eosin staining solution to prevent mold growth. It is also suitable for use in antimicrobial screening by the disk diffusion method.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-83-8

Specifications

Molecular Formula C10H14O
Quantity 500 g
InChI Key MGSRCZKZVOBKFT-UHFFFAOYSA-N
SMILES CC(C)C1=CC=C(C)C=C1O
IUPAC Name 5-methyl-2-(propan-2-yl)phenol
Molecular Weight (g/mol) 150.22
Product Type Stain Kit

The product is not intended for use as a biocide under global biocide regulations, including but not limited to US EPA's Federal Insecticide Fungicide and Rodenticide Act, European Biocidal Products Regulation, Canada's Pest Management Regulatory Agency, Turkey's Biocidal Products Regulation, Korea's Consumer Chemical Products and Biocide Safety Management Act (K-BPR) and others.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.