Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thymolphthalein TS/RS, 0.1% in Alcohol, Reagents

Catalog No. 01333445
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-445 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-445 Supplier Reagents Holdings Llc Supplier No. CT119500120A
Add to Cart
Add to Cart

Thymolphthalein TS/RS, 0.1% in Alcohol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 125-20-2
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym 5',5''-Diisopropyl-2',2''-dimethylphenolphthalein
IUPAC Name ethanol
SMILES CCO

Specifications

Boiling Point 78°C
Percent Purity 0.1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.8
Color Colorless
Quantity 120 mL
Physical Form Liquid
Chemical Name or Material Thymolphthalein TS/RS, 0.1% in Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.