Learn More
CPC Scientific H-Arg-Lys-Asp-Val-Tyr-OH 25MG
SDP
Supplier:  CPC Scientific THYM001B
Encompass
Thymopentin (TP5) is an active fragment of thymopoietin (TP) that reduces endocrine and behavioral responses to experimental stress. TP5 has immunomodulatory effects by suppression of proliferation of HL-60 cells.SEQUENCE: H-Arg-Lys-Asp-Val-Tyr-OHONE-LETTER SEQUENCE: RKDVYMOLECULAR FORMULA: C30H49N9O9MOLECULAR WEIGHT: 679.77STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [177966-81-3]SYNONYMS: Thymopoietin-5, Thymopoietin (32-36) (human, mouse), (Asp??)-Thymopoietin III (32-36) (bovine), TP-5, Thymopoietin I/II (32-36) (bovine)RESEARCH AREA: ImmunomodulationREFERENCES:G.Goldstein et al., Biol. Psychiatry, 48, 65 (2000)Y.-Z.Fan et al., Biochim. Biophys. Acta, 1763, 1059 (2006)SKU(s): THYM-001A, THYM-001B, THYM-001C
Catalog No. 50-211-0572
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.