Learn More
Bachem Thymosin b4 (human, bovine, horse, rat) Acetate
SDP
Supplier:  Bachem 4043020.0005
Encompass_Preferred
Thymosin b4 (human, bovine, horse, rat) Acetate, Ac-Ser-Asp-Lys-Pro-Asp-Met-Ala-Glu-Ile-Glu-Lys-Phe-Asp-Lys-Ser-Lys-Leu-Lys-Lys-Thr-Glu-Thr-Gln-Glu-Lys-Asn-Pro-Leu-Pro-Ser-Lys-Glu-Thr-Ile-Glu-Gln-Glu-Lys-Gln-Ala-Gly-Glu-Ser-OH, 77591-33-4, C212H350N56O78S, Mr 4963.51, 5mg. FX, Thymocrin. Thymosin b4 is a 43 amino acid peptide which is regarded as the main intracellular G-actin sequestering peptide. Extracellular thymosin b4 may contribute to physiological processes such as angiogenesis, wound healing, and regulation of inflammation. Additionally to its numerous functions thymosin b4 might also be of therapeutic value in the setting of acute myocardial damage.
Catalog No. 50-194-6818
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.