Learn More
CPC Scientific H-Lys-Leu-Lys-Lys-Thr-Glu-Thr-Gln-Glu-Lys-Asn-Pro-Leu-Pro-Ser-Lys-Glu-Thr-Ile-Glu-Gln-Glu-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific THYM005A
Encompass
Thymosin b4 fragment proven to restore the impaired T-lymphocyte blastogenic response of a uremic pneumonia patient.ONE-LETTER SEQUENCE: KLKKTETQEKNPLPSKETIEQEKMOLECULAR FORMULA: C118H204N32O41MOLECULAR WEIGHT:2727.07STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [113318-05-1], RESEARCH AREA: ImmunomodulationREFERENCES: T.Abiko and H.Sekino, Chem. Pharm. Bull., 35, 3757 (1987)
Catalog No. 50-211-0579
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.