Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Thyroglobulin Monoclonal Antibody (TG9.1), Invitrogen™

Catalog No. p-7245258
Change view
Click to view available options
Quantity:
1 mg
100 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546150 100 μg
01-671-549 500 μg
PIMA545995 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PIMA546150 Supplier Invitrogen™ Supplier No. MA546150
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-10596 detects Thyroglobulin, specifically T3, from human samples. MA1-10596 has been successfully used in ELISA applications. The MA1-10596 immunogen is purified human Thyroglobulin.

The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thyroglobulin
Applications ELISA
Classification Monoclonal
Clone TG9.1
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene TG
Gene Accession No. P01266
Gene Alias AITD3; cog; Colloid; congenital goiter; hereditary goitre; TG; Tgn; thyroglobulin; thyroglobulin precursor
Gene Symbols TG
Host Species Mouse
Immunogen Purified human Thyroglobulin.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7038
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.