Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thyroid Peroxidase Mouse anti-Human, HRP, Antibody Pair, Novus Biologicals™
SDP

Catalog No. p-7249240 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183332 5 Plates
NB183331 15 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB183332 Supplier Novus Biologicals Supplier No. NBP3067085PLATES
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antibody pair for Solid phase sandwich ELISA

Solid phase sandwich ELISA for quantitative determination of Human Thyroid Peroxidase.

Specifications

Antigen Thyroid Peroxidase
Applications ELISA
Classification Monoclonal
Conjugate HRP
Gene Alias EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8
Host Species Mouse
Quantity 5 Plates
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Gene ID (Entrez) 7173
Target Species Human
Content And Storage Storage of components varies. See protocol for specific instructions.
Product Type Antibody Pairs
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.