Learn More
Description
Specifications
Specifications
| Antigen | Thyroid Peroxidase |
| Applications | ELISA, Sandwich ELISA |
| Classification | Monoclonal |
| Conjugate | HRP |
| Gene Alias | EC 1.11.1, MSA, TDH2A, thyroid microsomal antigen, thyroid peroxidase, thyroperoxidase, TPXEC 1.11.1.8 |
| Host Species | Mouse |
| Quantity | 5 Plates |
| Regulatory Status | RUO |
| Research Discipline | Lipid and Metabolism |
| Gene ID (Entrez) | 7173 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
