Learn More
Carbosynth LLC. Rabbit polyclonal Thyroxine antibody
SDP
Supplier:  Carbosynth LLC. 20TR40
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
The thyroid hormones, thyroxine (T4) and triiodothyronine (T3), are tyrosine-based hormones produced by the thyroid gland primarily responsible for regulation of metabolism. An important component in the synthesis of thyroid hormones is iodine. The major form of thyroid hormone in the blood is thyroxine (T4), which has a longer half life than T3.
Catalog No. 50-125-2919
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.