Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier™ TI Light Probe
SDPSCI_ED

Catalog No. S16266ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
Elementary and Middle School
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S16266ND Elementary and Middle School
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16266ND Supplier Vernier™ Supplier No. TILTBTA

missing translation for 'validMainframeMsg'

Includes

TI Light Probe

Requires

Requires an interface and software, sold separately

Light probe by Texas Instruments

  • It senses both IR and visible light.
  • The reading reported is a relative number between 0 and 1.
  • Its low cost and ease of use make it an excellent choice for elementary and middle school.

Warranty and Services

5-year limited warranty

Specifications

Grade Elementary and Middle School
Interface Required
Warranty 5 Year Limited
For Use With (Application) Educational use only
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.