Learn More
Biocare Medical, LLC TIA-1 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM130C
Encompass_Preferred
TIA-1 antibody (T-cell intracytoplasmic antigen) is expressed in lymphocytes processing cytolytic potential. Studies show that 60 to 70% of anaplastic large cell lymphoma reacts with TIA-1. Studies also indicate that TIA-1 reacts with most large granular lymphocytic leukemias, hepatosplenic T-cell lymphomas, intestinal T-cell lymphomas, NK-like T-cell lymphomas, NK cell lymphomas, nasal T/NK-cell lymphomas, subcutaneous T-cell lymphomas and pulmonary angiocentric lymphomas of T-or NK-phenotype. All B-cell lymphomas, Hodgkin and lymphoblastic leukemias were negative for TIA-1.
Catalog No. 50-825-41
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.