Learn More
AdipoGen Tiadinil
SDP
Supplier:  AdipoGen CDXT0121M025
Encompass_Preferred
Chemical. CAS 223580-51-6. Formula C11H10ClN3OS. MW 267.7. Synthetic. Systemic anilide fungicide. Mode of action is through induc tion of plant defense mechanisms against a pathogen, therefore being a host plant defense inducer. Compound can be used as analytical reference material.
Catalog No. 50-255-9995
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.