Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tiagabine Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. T3165250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3165250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3165250MG Supplier TCI America Supplier No. T3165250MG

Chemical Identifiers

CAS 145821-59-6
Molecular Formula C20H26ClNO2S2
Molecular Weight (g/mol) 412.00
MDL Number MFCD07369025
InChI Key YUKARLAABCGMCN-PKLMIRHRSA-N
Synonym tiagabine hydrochloride, tiagabine hcl, gabitril, abbott 70569.hcl, unii-dqh6t6d8oy, dqh6t6d8oy, abbott-70569.hcl, r-1-4,4-bis 3-methylthiophen-2-yl but-3-en-1-yl piperidine-3-carboxylic acid hydrochloride, --r-1-4,4-bis 3-methyl-2-thienyl-3-butenyl nipecotic acid, hydrochloride
PubChem CID 91274
ChEBI CHEBI:85388
IUPAC Name hydrogen (3R)-1-[4,4-bis(3-methylthiophen-2-yl)but-3-en-1-yl]piperidine-3-carboxylic acid chloride
SMILES [H+].[Cl-].CC1=C(SC=C1)C(=CCCN1CCC[C@H](C1)C(O)=O)C1=C(C)C=CS1

Specifications

Melting Point 193°C
Color White-Yellow
Quantity 250 mg
Formula Weight 412.00
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tiagabine Hydrochloride
RTECSNumber TM6128245
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.