Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Basic Protection Polypropylene Gowns

Catalog No. 191303614 Shop All Tians International Products
Change view
Click to view available options
Color:
Blue
White
Yellow
Size:
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-130-3614 Blue X-Large
19-130-3612 White X-Large
19-130-3613 White 2X-Large
19-130-3610 Yellow X-Large
19-130-3611 Yellow 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-3614 Supplier Tians Supplier No. 813781XL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

This item is not returnable. View return policy

Low-lint polypropylene gowns for use in some critical environments, and many general application areas.

Ideal for environments where exposure to liquid contamination is not a concern.

  • Lightweight Full length
  • Elastic wrists

Specifications

Size X-Large
For Use With (Application) Critical Environments, General Industrial
No. of Pockets None
Gender Unisex
Packaging Type Cleanroom Packaged, 10 Pieces/Sealed Polybag, 5 Bags/Sealed Master Polybag
Product Type Controlled Environments Gown
Color Blue
Closure Type Neck and Waist Ties
Disposable Single-use
Sterility Non-sterile
Latex Free Yes
Sleeve Style Long Sleeve
Certifications/Compliance ISO Cleanroom Class 7
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.