Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians Environstar SPP Coveralls

Catalog No. 19063550 Shop All Tians International Products
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
4X-Large
Thickness:
Lightweight
Medium-weight
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness
19-063-550 3X-Large Lightweight
19-161-129C Large Medium-weight
19-161-129D X-Large Medium-weight
19-161-129F 3X-Large Medium-weight
19-161-129E 2X-Large -
19-161-129A Small -
19-161-129G 4X-Large Medium-weight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-063-550 Supplier Tians Supplier No. 21088XXXL
This item is not returnable. View return policy

For asbestos and abatement professionals

  • Polypropylene
  • Breathable
  • Collar
  • Elastic waist/back or elastic waist only
  • Hip pocket
  • White

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Material Polypropylene
Color White
Waist Style Elastic
Neck Style Collared
Thickness Lightweight
For Use With (Application) Asbestos and abatement professionals
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.