Learn More
Tians Environstar SPP Coveralls
Catalog No. 19161129A Shop All Tians International Products
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
3X-Large
4X-Large
Thickness:
Lightweight
Medium-weight
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness
19-161-129A Small -
19-161-129C Large Medium-weight
19-161-129D X-Large Medium-weight
19-161-129F 3X-Large Medium-weight
19-161-129E 2X-Large -
19-063-550 3X-Large Lightweight
19-161-129G 4X-Large Medium-weight
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-161-129A Supplier Tians Supplier No. 215883S
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
For asbestos and abatement professionals
  • Polypropylene
  • Breathable
  • Collar
  • Elastic waist/back or elastic waist only
  • Hip pocket
  • White
Specifications
Product Type Cleanroom Coveralls
Size Small
Material Polypropylene
Cuff Style Elastic