Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tians epic™ Lab Coats

Catalog No. 191301944B Shop All Tians International Products
Change view
Click to view available options
Color:
White
No. of Pockets:
1
Size:
Medium
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. of Pockets Size
19-130-1944B White 1 Medium
19-130-1944E White 1 2X-Large
19-130-1944C White 1 Large
19-130-1944D White 1 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1944B Supplier Tians Supplier No. 845881M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

Product Type Controlled Environments Lab Coat
Size Medium
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Knit
No. of Pockets 1
Certifications/Compliance ANSI, ASTM
For Use With (Application) For general protection and safety
Construction 1 oz. Fabric, Spunbond Polypropylene
Cleanroom Class Class 10,000 (ISO 7)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.