Learn More
Tians epic™ High Performance Static Dissipative Cleanroom Frocks
Shop All Tians International ProductsDescription
- Lint-free film coated frock
- Suitable for exposure to bloodborne pathogens, some hazardous liquids, and any wet general maintenance areas
- Features include a snap front, elastic wrists
- Mandarin snap collar, no pockets
- Fabric is constructed from a blend of polyethylene and polypropylene
- Finished fabric contains a proprietary additive making it static dissipative
- For use in ISO 6 Cleanroom Class environments
- Neatly folded and cleanroom packed
- 10 pieces per sealed inner polybag, 3 polybags per sealed master bag, 30 pieces per case
Specifications
Specifications
| Size | 2X-Large |
| Material | Polypropylene, Polyethylene |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Collar Type | Mandarin |
| Size (English) Chest | 11 in. |
| Size (Metric) Chest | 27.9 cm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.